siRNA Details
| Gene Symbol: LINC00899 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027036.1 | 1 | 249 | |||
| GenBank mRNA | AI760841.1 | - | 1 | 234 | ||
| AW137413.1 | - | 1 | 249 | |||
| CD674752.1 | - | 1 | 166 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00899| Gene Symbol: LINC00899 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027036.1 | 1 | 249 | |||
| GenBank mRNA | AI760841.1 | - | 1 | 234 | ||
| AW137413.1 | - | 1 | 249 | |||
| CD674752.1 | - | 1 | 166 | |||