siRNA Details
| Gene Symbol: LINC01341 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015422.1 | 2 | 518 | |||
| GenBank mRNA | AK022825.1 | - | 5 | 1958 | ||
| AK125177.1 | - | 2 | 1043 | |||
| BC101243.1 | - | 1 | 367 | |||
| BC101244.1 | - | 1 | 367 | |||
| BC101245.2 | - | 1 | 367 | |||
| BC101246.1 | - | 1 | 367 | |||

LINC01341
Genomic Map