siRNA Details
| Gene Symbol: SLC6A10P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003083.2 | 4, 5 | 2927 | |||
| GenBank mRNA | AK125843.1 | - | 4, 5 | 2928 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC6A10P| Gene Symbol: SLC6A10P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003083.2 | 4, 5 | 2927 | |||
| GenBank mRNA | AK125843.1 | - | 4, 5 | 2928 | ||