siRNA Details
| Gene Symbol: OR5E1P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_027711.1 | 1 | 459 | |||
| GenBank mRNA | AF309699.1 | - | LIT2079 OR5E1P. |
1 | 459 | |
| X89671.1 | - | 1 | 436 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR5E1P| Gene Symbol: OR5E1P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_027711.1 | 1 | 459 | |||
| GenBank mRNA | AF309699.1 | - | LIT2079 OR5E1P. |
1 | 459 | |
| X89671.1 | - | 1 | 436 | |||