siRNA Details
| Gene Symbol: FAM225A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK125961.1 | - | 1 | 3072 | ||
| CR627046.1 | - | 2 | 5938 | |||
| Gene Symbol: FAM225B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024376.1 | 3 | 5748 | |||
| GenBank mRNA | AL045730.1 | - | 1 | 497 | ||

FAM225A
Genomic Map