siRNA Details
| Gene Symbol: LINC00923 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024172.1 | 3 | 1327 | |||
| NR_024173.1 | 2 | 1155 | ||||
| GenBank mRNA | AK025311.1 | - | 3 | 1448 | ||
| BC105714.1 | - | 2 | 1155 | |||
| BC105715.1 | - | 3 | 1327 | |||
| BC105716.1 | - | 2 | 1155 | |||
| BC105717.1 | - | 2 | 1155 | |||

LINC00923
Genomic Map