siRNA Details
| Gene Symbol: DTYMK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033255.1 | 2, 3 | 420 | |||
| GenBank mRNA | BC171903.1 | - | 2, 3 | 359 | ||
| BG765702.1 | - | 2, 3 | 166 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DTYMK| Gene Symbol: DTYMK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033255.1 | 2, 3 | 420 | |||
| GenBank mRNA | BC171903.1 | - | 2, 3 | 359 | ||
| BG765702.1 | - | 2, 3 | 166 | |||