siRNA Details
| Gene Symbol: PAXIP1-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024476.1 | 4 | 1005 | |||
| NR_024477.1 | 4 | 1005 | ||||
| GenBank mRNA | AK056609.1 | - | 4 | 995 | ||
| BC033053.1 | - | 4 | 1031 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PAXIP1-AS2| Gene Symbol: PAXIP1-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024476.1 | 4 | 1005 | |||
| NR_024477.1 | 4 | 1005 | ||||
| GenBank mRNA | AK056609.1 | - | 4 | 995 | ||
| BC033053.1 | - | 4 | 1031 | |||