siRNA Details
| Gene Symbol: PAXIP1-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024476.1 | 3, 4 | 922 | |||
| NR_024477.1 | 3, 4 | 922 | ||||
| GenBank mRNA | AK056609.1 | - | 3, 4 | 912 | ||
| AK311721.1 | - | 2, 3 | 845 | |||
| BC033053.1 | - | 3, 4 | 948 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CA389913.1 | - | 3, 4 | 562 | Chr. 7 - 154928655 - 154946448 [+] on Build GRCh38 | |

PAXIP1-AS2
Genomic Map