siRNA Details
| Gene Symbol: CYP4F30P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_023391.1 | 5 | 2088 | |||
| GenBank mRNA | AL136789.1 | - | 5 | 2088 | ||
| Gene Symbol: CYP4F62P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103761.1 | 5 | 2261 | |||

CYP4F30P
Genomic Map