siRNA Details
| Gene Symbol: SNORD127 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003691.1 | 1 | 39 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SNORD127| Gene Symbol: SNORD127 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003691.1 | 1 | 39 | |||