siRNA Details
Gene Symbol: TIMM8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028383.1 | 1, 2 | 168 | |||
GenBank mRNA | AF165967.1 | - | 1, 2 | 78 | ||
BC000711.2 | - | 1, 2 | 115 | 2823930 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TIMM8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028383.1 | 1, 2 | 168 | |||
GenBank mRNA | AF165967.1 | - | 1, 2 | 78 | ||
BC000711.2 | - | 1, 2 | 115 | 2823930 |