siRNA Details
| Gene Symbol: TTTY14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001543.3 | 2 | 524 | |||
| NR_125733.1 | 4 | 718 | ||||
| NR_125737.1 | 3 | 159 | ||||
| GenBank mRNA | BG430850.1 | - | 5 | 356 | ||
| BM750145.1 | - | 3 | 160 | |||
| FY213451.1 | - | 4 | 381 | |||

TTTY14
Genomic Map