siRNA Details
| Gene Symbol: TTTY14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001543.3 | 2 | 559 | |||
| NR_125733.1 | 4 | 753 | ||||
| NR_125737.1 | 3 | 194 | ||||
| GenBank mRNA | AF332243.1 | - | 2 | 428 | ||
| BG430850.1 | - | 5 | 391 | |||
| BM750145.1 | - | 3 | 195 | |||
| FY213451.1 | - | 4 | 416 | |||

TTTY14
Genomic Map