siRNA Details
| Gene Symbol: TTTY14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001543.3 | 1 | 252 | |||
| NR_125733.1 | 1 | 252 | ||||
| NR_125734.1 | 1 | 252 | ||||
| NR_125735.1 | 1 | 252 | ||||
| GenBank mRNA | BI824313.1 | - | 1 | 46 | ||
| DA411985.1 | - | 1 | 252 | |||
| DB155066.1 | - | 1 | 245 | |||

TTTY14
Genomic Map