siRNA Details
| Gene Symbol: TTTY14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001543.3 | 1 | 149 | |||
| NR_125733.1 | 1 | 149 | ||||
| NR_125734.1 | 1 | 149 | ||||
| NR_125735.1 | 1 | 149 | ||||
| GenBank mRNA | DA411985.1 | - | 1 | 149 | ||
| DB155066.1 | - | 1 | 142 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AF332243.3 | - | 1 | 18 | Chr. Y - 18932699 - 19077416 [-] on Build GRCh38 | |

TTTY14
Genomic Map