siRNA Details
| Gene Symbol: GGNBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028361.1 | 1 | 181 | |||
| GenBank mRNA | AY383626.1 | - | 1 | 142 | ||
| AY383627.1 | - | 1 | 142 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GGNBP1| Gene Symbol: GGNBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028361.1 | 1 | 181 | |||
| GenBank mRNA | AY383626.1 | - | 1 | 142 | ||
| AY383627.1 | - | 1 | 142 | |||