siRNA Details
| Gene Symbol: LINC-PINT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015431.2 | 5 | 1841 | |||
| NR_109850.1 | 6 | 1965 | ||||
| NR_109851.1 | 6 | 1970 | ||||
| NR_109852.1 | 6 | 1943 | ||||
| GenBank mRNA | AA281849.1 | - | 1 | 92 | ||
| AK125651.1 | - | 6 | 1943 | |||
| AL713664.1 | - | 4 | 658 | |||
| BC047367.1 | - | 4 | 987 | |||
| BX451434.2 | - | 1 | 152 | |||

LINC-PINT
Genomic Map