siRNA Details
| Gene Symbol: SYT14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256006.1 | NP_001242935.1 | 3 | 330 | ||
| NR_027459.2 | 3 | 330 | ||||
| XM_017000932.1 | XP_016856421.1 | 3 | 320 | |||
| XM_017000933.1 | XP_016856422.1 | 3 | 183 | |||
| GenBank mRNA | AJ617624.1 | - | 2 | 211 | ||
| AJ617625.1 | - | 3 | 244 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DB084794.1 | - | 3 | 207 | Chr. 1 - 209938233 - 210015751 [+] on Build GRCh38 | |

SYT14
Genomic Map