siRNA Details
| Gene Symbol: SYT14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256006.1 | NP_001242935.1 | 2, 3 | 156 | ||
| NR_027459.2 | 2, 3 | 156 | ||||
| XM_017000932.1 | XP_016856421.1 | 2, 3 | 146 | |||
| GenBank mRNA | AJ617624.1 | - | 1, 2 | 37 | ||
| AJ617625.1 | - | 2, 3 | 70 | |||

SYT14
Genomic Map