siRNA Details
| Gene Symbol: FAM222A-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026661.2 | 4 | 826 | |||
| NR_026662.2 | 3 | 765 | ||||
| GenBank mRNA | BC007821.1 | - | 3 | 766 | ||
| BQ684893.1 | - | 3 | 787 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM222A-AS1| Gene Symbol: FAM222A-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026661.2 | 4 | 826 | |||
| NR_026662.2 | 3 | 765 | ||||
| GenBank mRNA | BC007821.1 | - | 3 | 766 | ||
| BQ684893.1 | - | 3 | 787 | |||