siRNA Details
| Gene Symbol: GUSBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003504.3 | 8 | 1037 | |||
| GenBank mRNA | BC034017.1 | - | 8 | 1038 | ||
| Gene Symbol: GUSBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132999.1 | 5 | 548 | |||
| GenBank mRNA | AL831827.1 | - | 1 | 926 | ||
| AW955360.1 | - | 4 | 376 | |||

GUSBP2
Genomic Map