siRNA Details
| Gene Symbol: GUSBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003504.3 | 7, 8 | 977 | |||
| GenBank mRNA | BC034017.1 | - | 7, 8 | 978 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GUSBP2| Gene Symbol: GUSBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003504.3 | 7, 8 | 977 | |||
| GenBank mRNA | BC034017.1 | - | 7, 8 | 978 | ||