siRNA Details
| Gene Symbol: SMA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034021.1 | 2 | 147 | |||
| GenBank mRNA | X83301.1 | - | 2 | 147 | ||
| Gene Symbol: GUSBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003504.3 | 2 | 209 | |||
| GenBank mRNA | BC034017.1 | - | 2 | 210 | ||
| BC140728.1 | - | 1 | 32 | |||
| BC171739.1 | - | 1 | 32 | 9052954 | ||

SMA5
Genomic Map