siRNA Details
| Gene Symbol: LOC100132831 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027761.1 | 1 | 1161 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100132831| Gene Symbol: LOC100132831 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027761.1 | 1 | 1161 | |||