siRNA Details
| Gene Symbol: RACGAP1P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026583.1 | 1 | 2405 | |||
| GenBank mRNA | AF334184.1 | - | 1 | 2413 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RACGAP1P| Gene Symbol: RACGAP1P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026583.1 | 1 | 2405 | |||
| GenBank mRNA | AF334184.1 | - | 1 | 2413 | ||