siRNA Details
Gene Symbol: HCG9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028032.1 | 1 | 60 | |||
GenBank mRNA | AK289926.1 | - | 1 | 83 | ||
DA293452.1 | - | 1 | 83 | |||
X95289.1 | - | 1 | 60 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: HCG9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028032.1 | 1 | 60 | |||
GenBank mRNA | AK289926.1 | - | 1 | 83 | ||
DA293452.1 | - | 1 | 83 | |||
X95289.1 | - | 1 | 60 |