siRNA Details
| Gene Symbol: KHDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001251874.1 | NP_001238803.1 | 1 | 538 | ||
| NR_027005.2 | 2 | 111 | ||||
| GenBank mRNA | AK090984.1 | - | 1 | 438 | ||
| BM747426.1 | - | 2 | 111 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KHDC1| Gene Symbol: KHDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001251874.1 | NP_001238803.1 | 1 | 538 | ||
| NR_027005.2 | 2 | 111 | ||||
| GenBank mRNA | AK090984.1 | - | 1 | 438 | ||
| BM747426.1 | - | 2 | 111 | |||