siRNA Details
| Gene Symbol: PIGP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016430.3 | NP_057514.2 | 3 | 316 | ||
| XM_017028365.1 | XP_016883854.1 | 3 | 258 | |||
| GenBank mRNA | AB037164.1 | - | 2 | 167 | ||
| AF216305.1 | - | 3 | 232 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PIGP| Gene Symbol: PIGP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016430.3 | NP_057514.2 | 3 | 316 | ||
| XM_017028365.1 | XP_016883854.1 | 3 | 258 | |||
| GenBank mRNA | AB037164.1 | - | 2 | 167 | ||
| AF216305.1 | - | 3 | 232 | |||