siRNA Details
| Gene Symbol: TDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001578.1 | 4 | 373 | |||
| GenBank mRNA | AJ301562.1 | - | 4 | 373 | ||
| AK057762.1 | - | 5 | 462 | |||
| AY101186.1 | - | 3 | 275 | |||
| AY101187.1 | - | 3 | 275 | |||
| BC074983.2 | - | 4 | 310 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX091784.1 | - | 4 | 328 | Chr. 8 - 11346178 - 11361742 [+] on Build GRCh38 | |

TDH
Genomic Map