siRNA Details
| Gene Symbol: TDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001578.1 | 8 | 1089 | |||
| GenBank mRNA | AJ301562.1 | - | 8 | 1089 | ||
| AK057762.1 | - | 8 | 1028 | |||
| AY101186.1 | - | 7 | 991 | |||
| AY101187.1 | - | 7 | 907 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TDH| Gene Symbol: TDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001578.1 | 8 | 1089 | |||
| GenBank mRNA | AJ301562.1 | - | 8 | 1089 | ||
| AK057762.1 | - | 8 | 1028 | |||
| AY101186.1 | - | 7 | 991 | |||
| AY101187.1 | - | 7 | 907 | |||