siRNA Details
| Gene Symbol: TDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001578.1 | 7 | 791 | |||
| GenBank mRNA | AJ301562.1 | - | 7 | 791 | ||
| AK057762.1 | - | 7 | 730 | |||
| AY101186.1 | - | 6 | 693 | |||
| AY101187.1 | - | 6 | 609 | |||
| BC074983.2 | - | 6 | 578 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | HY113237.1 | - | 3 | 322 | Chr. 8 - 11361473 - 11365598 [+] on Build GRCh38 | |

TDH
Genomic Map