siRNA Details
| Gene Symbol: UBAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033243.2 | 1, 2 | 244 | |||
| GenBank mRNA | BP237024.1 | - | 1, 2 | 212 | ||
| BX473128.1 | - | 1, 2 | 236 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

UBAP1| Gene Symbol: UBAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033243.2 | 1, 2 | 244 | |||
| GenBank mRNA | BP237024.1 | - | 1, 2 | 212 | ||
| BX473128.1 | - | 1, 2 | 236 | |||