siRNA Details
| Gene Symbol: CLEC4M | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026707.1 | 4 | 1107 | |||
| NR_026708.1 | 4 | 1107 | ||||
| NR_026709.1 | 3 | 1044 | ||||
| GenBank mRNA | AK023750.1 | - | 2 | 828 | ||
| AY042235.1 | - | 4 | 989 | |||
| AY042236.1 | - | 4 | 989 | |||
| AY042239.1 | - | 3 | 926 | |||

CLEC4M
Genomic Map