siRNA Details
| Gene Symbol: FAM186B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027450.1 | 1 | 357 | |||
| XM_017020008.1 | XP_016875497.1 | 1 | 440 | |||
| GenBank mRNA | DC402111.1 | - | 1 | 348 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DB446089.1 | - | 1 | 332 | Chr. 12 - 49604482 - 49605602 [-] on Build GRCh38 | |

FAM186B
Genomic Map