siRNA Details
| Gene Symbol: FAM186B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027450.1 | 7 | 2996 | |||
| XM_006719625.2 | XP_006719688.1 | 8 | 3019 | |||
| XM_006719626.2 | XP_006719689.1 | 7 | 2828 | |||
| XM_017020008.1 | XP_016875497.1 | 8 | 3270 | |||
| GenBank mRNA | AK302702.1 | - | 7 | 2996 | ||

FAM186B
Genomic Map