siRNA Details
| Gene Symbol: FAM186B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027450.1 | 1 | 265 | |||
| XM_017020008.1 | XP_016875497.1 | 1 | 348 | |||
| GenBank mRNA | AK302702.1 | - | 1 | 265 | ||
| DC402111.1 | - | 1 | 256 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM186B| Gene Symbol: FAM186B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027450.1 | 1 | 265 | |||
| XM_017020008.1 | XP_016875497.1 | 1 | 348 | |||
| GenBank mRNA | AK302702.1 | - | 1 | 265 | ||
| DC402111.1 | - | 1 | 256 | |||