siRNA Details
| Gene Symbol: LINC00710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015413.1 | 6 | 737 | |||
| NR_024410.1 | 5 | 667 | ||||
| NR_024411.1 | 4 | 556 | ||||
| GenBank mRNA | BC033338.1 | - | 6 | 669 | ||
| BC106956.2 | - | 5 | 471 | |||
| BC106957.2 | - | 4 | 360 | |||
| BC113009.1 | - | 4 | 360 | |||

LINC00710
Genomic Map