siRNA Details
| Gene Symbol: LINC00710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015413.1 | 6 | 916 | |||
| NR_024410.1 | 5 | 846 | ||||
| NR_024411.1 | 4 | 735 | ||||
| GenBank mRNA | BC033338.1 | - | 6 | 848 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DB508949.2 | - | 3 | 490 | Chr. 10 - 10934950 - 10946576 [-] on Build GRCh38 | |

LINC00710
Genomic Map