siRNA Details
| Gene Symbol: RNU105B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_004386.2 | 1 | 51 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RNU105B| Gene Symbol: RNU105B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_004386.2 | 1 | 51 | |||