siRNA Details
| Gene Symbol: PSMA3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029434.1 | 5 | 1520 | |||
| NR_029435.1 | 4 | 1479 | ||||
| GenBank mRNA | BC034618.1 | - | 5 | 1512 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PSMA3-AS1| Gene Symbol: PSMA3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029434.1 | 5 | 1520 | |||
| NR_029435.1 | 4 | 1479 | ||||
| GenBank mRNA | BC034618.1 | - | 5 | 1512 | ||