siRNA Details
Gene Symbol: PSMA3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029434.1 | 5 | 902 | |||
NR_029435.1 | 4 | 861 | ||||
GenBank mRNA | BC034618.1 | - | 5 | 894 | ||
BX460997.2 | - | 4 | 845 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: PSMA3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029434.1 | 5 | 902 | |||
NR_029435.1 | 4 | 861 | ||||
GenBank mRNA | BC034618.1 | - | 5 | 894 | ||
BX460997.2 | - | 4 | 845 |