siRNA Details
| Gene Symbol: SNORA72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002581.1 | 1 | 52 | |||
| GenBank mRNA | Y11167.1 | - | 1 | 52 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK128768.1 | - | 2 | 1551 | Chr. 8 - 98041726 - 98044121 [-] on Build GRCh38 | |

SNORA72
Genomic Map