siRNA Details
| Gene Symbol: SNORA70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_000011.1 | 1 | 16 | |||
| GenBank mRNA | Y11164.1 | - | 1 | 16 | ||
| Gene Symbol: RPL10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF218023.1 | - | 3 | 1596 | ||
| AK027197.1 | - | 1 | 1821 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CR610716.1 | - | 2 | 1005 | Chr. X - 154398496 - 154400312 [+] on Build GRCh38 | |

SNORA70
Genomic Map