siRNA Details
| Gene Symbol: SNORA69 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002584.1 | 1 | 117 | |||
| GenBank mRNA | Y11163.1 | - | 1 | 117 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX421532.2 | - | 1 | 519 | Chr. X - 119787067 - 119787886 [-] on Build GRCh38 | |

SNORA69
Genomic Map