siRNA Details
| Gene Symbol: FAM95B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026759.1 | 1 | 416 | |||
| GenBank mRNA | AK125850.1 | - | 1 | 2688 | ||
| Gene Symbol: LOC105379252 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136310.1 | 1 | 416 | |||

FAM95B1
Genomic Map