siRNA Details
| Gene Symbol: FAM95B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026759.1 | 3 | 794 | |||
| GenBank mRNA | AL833349.1 | - | 1 | 386 | ||
| Gene Symbol: LOC105379252 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136310.1 | 3 | 794 | |||

FAM95B1
Genomic Map