siRNA Details
| Gene Symbol: SEC61A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024576.2 | 12 | 1507 | |||
| NR_024577.2 | 13 | 1640 | ||||
| GenBank mRNA | AK057532.1 | - | 13 | 1581 | ||
| BC026179.1 | - | 12 | 1457 | 4823130 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SEC61A2| Gene Symbol: SEC61A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024576.2 | 12 | 1507 | |||
| NR_024577.2 | 13 | 1640 | ||||
| GenBank mRNA | AK057532.1 | - | 13 | 1581 | ||
| BC026179.1 | - | 12 | 1457 | 4823130 | ||