siRNA Details
Gene Symbol: LINC00326 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026969.1 | 1 | 259 | |||
GenBank mRNA | BC039371.1 | - | 1 | 259 | ||
BI460768.1 | - | 1 | 219 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00326 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026969.1 | 1 | 259 | |||
GenBank mRNA | BC039371.1 | - | 1 | 259 | ||
BI460768.1 | - | 1 | 219 |