siRNA Details
Gene Symbol: GNAS-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002785.2 | 4 | 807 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000424094.6 | - | 4 | 807 | Chr. 20 - 58818919 - 58850903 [-] on Build GRCh38 |
